Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACACGGTGCTCAAGATCACGGACTT[C/T]GGCCTCGCCCGCGAGTGGCACAAGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600137 | ||||||||||||||||||||
Literature Links: |
MAP3K10 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MAP3K10 - mitogen-activated protein kinase kinase kinase 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002446.3 | 1035 | Silent Mutation | TTC,TTT | F,F 249 | NP_002437.2 | |
XM_006723222.2 | 1035 | UTR 5 | XP_006723285.1 | |||
XM_006723223.3 | 1035 | Intron | XP_006723286.1 | |||
XM_011526981.2 | 1035 | Silent Mutation | TTC,TTT | F,F 249 | XP_011525283.1 | |
XM_011526982.2 | 1035 | Silent Mutation | TTC,TTT | F,F 249 | XP_011525284.1 |