Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTCTGCCCTCCTACTCCCCAGGCT[A/G]TGATCCAAAATCCCTTCAGCAATGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616582 MIM: 600390 | ||||||||||||||||||||
Literature Links: |
LSR PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LSR - lipolysis stimulated lipoprotein receptor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
USF2 - upstream transcription factor 2, c-fos interacting | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321150.1 | 456 | Intron | NP_001308079.1 | |||
NM_003367.3 | 456 | Missense Mutation | ATG,GTG | M,V 145 | NP_003358.1 | |
NM_207291.2 | 456 | Missense Mutation | ATG,GTG | M,V 78 | NP_997174.1 | |
XM_005259197.3 | 456 | Missense Mutation | ATG,GTG | M,V 145 | XP_005259254.1 | |
XM_011527260.2 | 456 | Missense Mutation | ATG,GTG | M,V 145 | XP_011525562.1 | |
XM_011527261.2 | 456 | Missense Mutation | ATG,GTG | M,V 145 | XP_011525563.1 | |
XM_017027199.1 | 456 | Missense Mutation | ATG,GTG | M,V 78 | XP_016882688.1 |