Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTCTCCCCTGGCAGAGCAGAGGAA[G/T]CGTTACTCCACAGTTGTTATGGCTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 614987 | ||||||||||||||||||||
Literature Links: |
EPS8L1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
EPS8L1 - EPS8 like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017729.3 | 173 | Intron | NP_060199.3 | |||
NM_133180.2 | 173 | Missense Mutation | AAG,AAT | K,N 23 | NP_573441.2 | |
XM_005259020.1 | 173 | Missense Mutation | AAG,AAT | K,N 23 | XP_005259077.1 | |
XM_011527050.1 | 173 | Intron | XP_011525352.1 | |||
XM_011527051.2 | 173 | Missense Mutation | AAG,AAT | K,N 23 | XP_011525353.1 | |
XM_011527052.2 | 173 | Missense Mutation | AAG,AAT | K,N 23 | XP_011525354.1 |
RDH13 - retinol dehydrogenase 13 (all-trans/9-cis) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |