Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATAAAGTTCACAACCTTCACTGAAT[G/T]GTGTCAACCTGTACAGAGAATCATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZFP30 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZFP30 - ZFP30 zinc finger protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320666.1 | 2427 | UTR 3 | NP_001307595.1 | |||
NM_001320667.1 | 2427 | UTR 3 | NP_001307596.1 | |||
NM_001320668.1 | 2427 | UTR 3 | NP_001307597.1 | |||
NM_001320669.1 | 2427 | UTR 3 | NP_001307598.1 | |||
NM_001320670.1 | 2427 | UTR 3 | NP_001307599.1 | |||
NM_001320671.1 | 2427 | UTR 3 | NP_001307600.1 | |||
NM_014898.2 | 2427 | UTR 3 | NP_055713.1 |