Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGTGAGCAGTCCGGGCCCGGTCCT[G/T]GAGGGGGCTGAGGCTGCCGTCGTGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608153 | ||||||||||||||||||||
Literature Links: |
PPP1R14A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PPP1R14A - protein phosphatase 1 regulatory inhibitor subunit 14A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243947.1 | 667 | Missense Mutation | AAG,CAG | K,Q 112 | NP_001230876.1 | |
NM_033256.2 | 667 | Missense Mutation | AAG,CAG | K,Q 139 | NP_150281.1 |