Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTCAGTGTGGCGTTAGCGATGCTT[C/T]TTCATGGTGTCATTTATCCACTTGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602652 MIM: 604438 | ||||||||||||||||||||
Literature Links: |
KLK6 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KLK6 - kallikrein related peptidase 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
KLK7 - kallikrein related peptidase 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001207053.1 | 1022 | Silent Mutation | AAA,AAG | K,K 178 | NP_001193982.1 | |
NM_001243126.1 | 1022 | Silent Mutation | AAA,AAG | K,K 243 | NP_001230055.1 | |
NM_005046.3 | 1022 | Silent Mutation | AAA,AAG | K,K 250 | NP_005037.1 | |
NM_139277.2 | 1022 | Silent Mutation | AAA,AAG | K,K 250 | NP_644806.1 |