Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCACGATGCTAAGCGTGATGAAGA[C/T]GCCCGTCCTGCCCACGCCGGCACTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601576 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PTPRS PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PTPRS - protein tyrosine phosphatase, receptor type S | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002850.3 | 5828 | Missense Mutation | ATC,GTC | I,V 1889 | NP_002841.3 | |
NM_130853.2 | 5828 | Missense Mutation | ATC,GTC | I,V 1442 | NP_570923.2 | |
NM_130854.2 | 5828 | Missense Mutation | ATC,GTC | I,V 1851 | NP_570924.2 | |
NM_130855.2 | 5828 | Missense Mutation | ATC,GTC | I,V 1446 | NP_570925.2 | |
XM_005259600.2 | 5828 | Missense Mutation | ATC,GTC | I,V 1876 | XP_005259657.1 | |
XM_005259606.2 | 5828 | Missense Mutation | ATC,GTC | I,V 1851 | XP_005259663.1 | |
XM_005259607.2 | 5828 | Missense Mutation | ATC,GTC | I,V 1847 | XP_005259664.1 | |
XM_005259609.1 | 5828 | Missense Mutation | ATC,GTC | I,V 1442 | XP_005259666.1 | |
XM_005259610.1 | 5828 | Missense Mutation | ATC,GTC | I,V 1438 | XP_005259667.1 | |
XM_011528157.2 | 5828 | Missense Mutation | ATC,GTC | I,V 1458 | XP_011526459.1 | |
XM_011528158.2 | 5828 | Missense Mutation | ATC,GTC | I,V 1098 | XP_011526460.1 | |
XM_017027065.1 | 5828 | Missense Mutation | ATC,GTC | I,V 1880 | XP_016882554.1 | |
XM_017027066.1 | 5828 | Missense Mutation | ATC,GTC | I,V 1880 | XP_016882555.1 | |
XM_017027067.1 | 5828 | Missense Mutation | ATC,GTC | I,V 1880 | XP_016882556.1 | |
XM_017027068.1 | 5828 | Missense Mutation | ATC,GTC | I,V 1876 | XP_016882557.1 | |
XM_017027069.1 | 5828 | Missense Mutation | ATC,GTC | I,V 1871 | XP_016882558.1 | |
XM_017027070.1 | 5828 | Missense Mutation | ATC,GTC | I,V 1864 | XP_016882559.1 | |
XM_017027071.1 | 5828 | Missense Mutation | ATC,GTC | I,V 1860 | XP_016882560.1 | |
XM_017027072.1 | 5828 | Missense Mutation | ATC,GTC | I,V 1851 | XP_016882561.1 | |
XM_017027073.1 | 5828 | Missense Mutation | ATC,GTC | I,V 1638 | XP_016882562.1 | |
XM_017027074.1 | 5828 | Missense Mutation | ATC,GTC | I,V 1462 | XP_016882563.1 | |
XM_017027075.1 | 5828 | Missense Mutation | ATC,GTC | I,V 1458 | XP_016882564.1 | |
XM_017027076.1 | 5828 | Missense Mutation | ATC,GTC | I,V 1446 | XP_016882565.1 |