Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGAACCAAAAACACAGAGATGAG[C/T]GTCATCAAGACGGGAATGACTACGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604144 | ||||||||||||||||||||
Literature Links: |
SLC7A9 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SLC7A9 - solute carrier family 7 member 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001126335.1 | 1371 | Silent Mutation | ACA,ACG | T,T 418 | NP_001119807.1 | |
NM_001243036.1 | 1371 | Silent Mutation | ACA,ACG | T,T 418 | NP_001229965.1 | |
NM_014270.4 | 1371 | Silent Mutation | ACA,ACG | T,T 418 | NP_055085.1 | |
XM_006722992.1 | 1371 | Silent Mutation | ACA,ACG | T,T 191 | XP_006723055.1 | |
XM_011526402.2 | 1371 | Silent Mutation | ACA,ACG | T,T 418 | XP_011524704.1 | |
XM_017026230.1 | 1371 | Silent Mutation | ACA,ACG | T,T 330 | XP_016881719.1 |
TDRD12 - tudor domain containing 12 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |