Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAGTTTCCCCACATTCTTTACATT[C/T]ATAGGACTTTTCACCAGCATGAGTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ZNF607 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF607 - zinc finger protein 607 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001172677.1 | 2232 | Missense Mutation | AAA,GAA | K,E 589 | NP_001166148.1 | |
NM_032689.4 | 2232 | Missense Mutation | AAA,GAA | K,E 590 | NP_116078.4 | |
XM_006723435.3 | 2232 | Missense Mutation | AAA,GAA | K,E 590 | XP_006723498.1 | |
XM_006723436.2 | 2232 | Missense Mutation | AAA,GAA | K,E 589 | XP_006723499.1 | |
XM_006723437.2 | 2232 | Missense Mutation | AAA,GAA | K,E 557 | XP_006723500.1 | |
XM_006723440.1 | 2232 | Missense Mutation | AAA,GAA | K,E 499 | XP_006723503.1 | |
XM_011527414.2 | 2232 | Missense Mutation | AAA,GAA | K,E 499 | XP_011525716.1 |
ZNF781 - zinc finger protein 781 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |