Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAAGGAGCCAAATTAGGCTTCTGT[G/T]TCCCTGATCTGGCACTCCAAGGGGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 124089 MIM: 609358 MIM: 613232 | ||||||||||||||||||||
Literature Links: |
COX6B1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
COX6B1 - cytochrome c oxidase subunit 6B1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ETV2 - ETS variant 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300974.1 | 1006 | UTR 5 | NP_001287903.1 | |||
NM_001304549.1 | 1006 | Missense Mutation | GTC,TTC | V,F 32 | NP_001291478.1 | |
NM_014209.3 | 1006 | Missense Mutation | GTC,TTC | V,F 32 | NP_055024.2 | |
XM_005258652.2 | 1006 | Missense Mutation | GTC,TTC | V,F 60 | XP_005258709.1 | |
XM_011526624.2 | 1006 | UTR 5 | XP_011524926.1 | |||
XM_017026472.1 | 1006 | Missense Mutation | GTC,TTC | V,F 60 | XP_016881961.1 |
RBM42 - RNA binding motif protein 42 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |