Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCATGCTTCCCAACCCCAGGGCCAC[A/G]CACGCACCATAAATTTTAAAAGCAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610645 MIM: 608349 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CIB3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CIB3 - calcium and integrin binding family member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300922.1 | Intron | NP_001287851.1 | ||||
NM_054113.3 | Intron | NP_473454.1 |
HSH2D - hematopoietic SH2 domain containing | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |