Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGATGTGGGCTTTGATGGAGGCAC[C/T]GATCGTATCTTCCTCGTGTCCCCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602488 MIM: 603953 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CYTH2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CYTH2 - cytohesin 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
KCNJ14 - potassium voltage-gated channel subfamily J member 14 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_013348.3 | 996 | Silent Mutation | ACC,ACT | T,T 263 | NP_037480.1 |
LOC105372430 - uncharacterized LOC105372430 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |