Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGACCCATGATGACTTCTCCACTCA[C/T]ACGGCTTTCATATGGACCTACTCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF799 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF799 - zinc finger protein 799 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080821.2 | 973 | Missense Mutation | ATG,GTG | M,V 107 | NP_001074290.1 | |
NM_001322497.1 | 973 | Missense Mutation | ATG,GTG | M,V 75 | NP_001309426.1 | |
NM_001322498.1 | 973 | Missense Mutation | ATG,GTG | M,V 75 | NP_001309427.1 | |
XM_017027452.1 | 973 | Missense Mutation | ATG,GTG | M,V 75 | XP_016882941.1 |