Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGTTCCCGATGACCTTGGGCAGG[C/T]GCTGCAGTGTTCCTACATCGGCAGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600696 MIM: 602518 | ||||||||||||||||||||
Literature Links: |
ECH1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ECH1 - enoyl-CoA hydratase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001398.2 | 717 | Missense Mutation | CCC,CTC | P,L 211 | NP_001389.2 | |
XM_017026448.1 | 717 | Missense Mutation | CCC,CTC | P,L 131 | XP_016881937.1 |
LGALS4 - galectin 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |