Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCCCCACCTATCCCCAGAAACCCC[A/C]GCCCCGGATGGCCCCCGGTATCGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605248 MIM: 603197 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MCOLN1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MCOLN1 - mucolipin 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PNPLA6 - patatin like phospholipase domain containing 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001166111.1 | 394 | Silent Mutation | CCA,CCC | P,P 89 | NP_001159583.1 | |
NM_001166112.1 | 394 | Silent Mutation | CCA,CCC | P,P 41 | NP_001159584.1 | |
NM_001166113.1 | 394 | Silent Mutation | CCA,CCC | P,P 41 | NP_001159585.1 | |
NM_001166114.1 | 394 | Silent Mutation | CCA,CCC | P,P 80 | NP_001159586.1 | |
NM_006702.4 | 394 | Silent Mutation | CCA,CCC | P,P 41 | NP_006693.3 |