Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATGCTCACTCACCTGAGAGGCTGA[A/G]CCCCTGACAGCGTTTGCGTCCTCCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606094 | ||||||||||||||||||||
Literature Links: |
CEACAM18 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CEACAM18 - carcinoembryonic antigen related cell adhesion molecule 18 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SIGLEC12 - sialic acid binding Ig like lectin 12 (gene/pseudogene) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_033329.2 | Intron | NP_201586.1 | ||||
NM_053003.3 | Intron | NP_443729.1 | ||||
XM_011527441.2 | Intron | XP_011525743.1 | ||||
XM_011527442.2 | Intron | XP_011525744.1 |