Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAGCCAGGCCTCAGTGAGGTCATC[A/G]GTGGTGCGGATTTCGAACACCTGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604104 MIM: 612807 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GMFG PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GMFG - glia maturation factor gamma | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301008.1 | 488 | Silent Mutation | ACC,ACT | T,T 85 | NP_001287937.1 | |
NM_004877.3 | 488 | Silent Mutation | ACC,ACT | T,T 126 | NP_004868.1 |
LRFN1 - leucine rich repeat and fibronectin type III domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |