Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGTAGGAAGCAGTGACCTTCAAGGA[C/T]GTGGCTGTGGCCTTCACGGAGGAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF226 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF226 - zinc finger protein 226 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001032372.1 | 341 | Silent Mutation | GAC,GAT | D,D 12 | NP_001027544.1 | |
NM_001032373.1 | 341 | Silent Mutation | GAC,GAT | D,D 12 | NP_001027545.1 | |
NM_001032374.1 | 341 | Silent Mutation | GAC,GAT | D,D 12 | NP_001027546.1 | |
NM_001146220.2 | 341 | Silent Mutation | GAC,GAT | D,D 12 | NP_001139692.1 | |
NM_001319088.1 | 341 | Silent Mutation | GAC,GAT | D,D 12 | NP_001306017.1 | |
NM_001319089.1 | 341 | Silent Mutation | GAC,GAT | D,D 12 | NP_001306018.1 | |
NM_001319090.1 | 341 | Silent Mutation | GAC,GAT | D,D 12 | NP_001306019.1 | |
NM_015919.3 | 341 | Silent Mutation | GAC,GAT | D,D 12 | NP_057003.2 | |
NM_016444.2 | 341 | Silent Mutation | GAC,GAT | D,D 12 | NP_057528.2 | |
XM_005259227.2 | 341 | Silent Mutation | GAC,GAT | D,D 12 | XP_005259284.1 | |
XM_006723367.3 | 341 | Silent Mutation | GAC,GAT | D,D 12 | XP_006723430.1 | |
XM_006723368.3 | 341 | Silent Mutation | GAC,GAT | D,D 12 | XP_006723431.1 | |
XM_006723369.2 | 341 | Silent Mutation | GAC,GAT | D,D 12 | XP_006723432.1 | |
XM_017027262.1 | 341 | Silent Mutation | GAC,GAT | D,D 12 | XP_016882751.1 | |
XM_017027263.1 | 341 | UTR 5 | XP_016882752.1 | |||
XM_017027264.1 | 341 | UTR 5 | XP_016882753.1 | |||
XM_017027265.1 | 341 | Silent Mutation | GAC,GAT | D,D 12 | XP_016882754.1 |