Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCCAGCCGGTAGGTTCCAGGCCCC[A/G]GCCGCCTCCGCCCCAGCTGCAGGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602565 MIM: 608529 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCL25 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCL25 - C-C motif chemokine ligand 25 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
FBN3 - fibrillin 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321431.1 | 8443 | Missense Mutation | CCG,CTG | P,L 2768 | NP_001308360.1 | |
NM_032447.4 | 8443 | Missense Mutation | CCG,CTG | P,L 2768 | NP_115823.3 | |
XM_017027372.1 | 8443 | Missense Mutation | CCG,CTG | P,L 2768 | XP_016882861.1 | |
XM_017027373.1 | 8443 | Missense Mutation | CCG,CTG | P,L 2748 | XP_016882862.1 | |
XM_017027374.1 | 8443 | Missense Mutation | CCG,CTG | P,L 2736 | XP_016882863.1 | |
XM_017027375.1 | 8443 | Missense Mutation | CCG,CTG | P,L 2727 | XP_016882864.1 | |
XM_017027376.1 | 8443 | Missense Mutation | CCG,CTG | P,L 2725 | XP_016882865.1 | |
XM_017027377.1 | 8443 | Missense Mutation | CCG,CTG | P,L 1907 | XP_016882866.1 | |
XM_017027378.1 | 8443 | Missense Mutation | CCG,CTG | P,L 1554 | XP_016882867.1 | |
XM_017027379.1 | 8443 | Missense Mutation | CCG,CTG | P,L 2810 | XP_016882868.1 |