Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGGGTCTACTATTGCCTGGTGAA[C/T]ATCACGCTGAGTGACCTGCTCACGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609156 MIM: 603751 | ||||||||||||||||||||
Literature Links: |
NCLN PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NCLN - nicalin | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
S1PR4 - sphingosine-1-phosphate receptor 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003775.3 | 316 | Silent Mutation | AAC,AAT | N,N 87 | NP_003766.1 |