Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCACCACACGGGTGGGCTGTGTGC[A/G]TCCACAGGTATGACAGGTGCGAGGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600128 | ||||||||||||||||||||
Literature Links: |
KIAA1683 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KIAA1683 - KIAA1683 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145304.1 | 3997 | Missense Mutation | NP_001138776.1 | |||
NM_001145305.1 | 3997 | Missense Mutation | NP_001138777.1 | |||
NM_025249.3 | 3997 | Missense Mutation | NP_079525.1 | |||
XM_005260084.1 | 3997 | Missense Mutation | XP_005260141.1 | |||
XM_011528324.1 | 3997 | Missense Mutation | XP_011526626.1 |
LOC729966 - uncharacterized LOC729966 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PDE4C - phosphodiesterase 4C | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |