Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCTCAGAGGAAGTGTTGGTTTCTA[C/T]TTAAGAACATAGCAATGTTAAAGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612536 MIM: 613583 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
THAP8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
THAP8 - THAP domain containing 8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
WDR62 - WD repeat domain 62 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001083961.1 | Intron | NP_001077430.1 | ||||
NM_173636.4 | Intron | NP_775907.4 | ||||
XM_005258809.2 | Intron | XP_005258866.1 | ||||
XM_011526837.1 | Intron | XP_011525139.1 | ||||
XM_011526838.1 | Intron | XP_011525140.1 | ||||
XM_011526839.1 | Intron | XP_011525141.1 | ||||
XM_011526840.2 | Intron | XP_011525142.1 | ||||
XM_011526841.2 | Intron | XP_011525143.1 | ||||
XM_011526842.1 | Intron | XP_011525144.1 | ||||
XM_011526843.1 | Intron | XP_011525145.1 | ||||
XM_011526844.2 | Intron | XP_011525146.1 | ||||
XM_017026665.1 | Intron | XP_016882154.1 |