Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCTTCCTTTCTCCTCTAGGGCCCA[C/T]CAGCTCTGAGCAGATCATGAAGACA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600040 MIM: 134790 | ||||||||||||||||||||
Literature Links: |
BAX PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
BAX - BCL2 associated X, apoptosis regulator | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001291428.1 | 110 | Missense Mutation | ACC,ATC | T,I 14 | NP_001278357.1 | |
NM_001291429.1 | 110 | Intron | NP_001278358.1 | |||
NM_001291430.1 | 110 | Intron | NP_001278359.1 | |||
NM_001291431.1 | 110 | Intron | NP_001278360.1 | |||
NM_004324.3 | 110 | Missense Mutation | ACC,ATC | T,I 14 | NP_004315.1 | |
NM_138761.3 | 110 | Missense Mutation | ACC,ATC | T,I 14 | NP_620116.1 | |
NM_138763.3 | 110 | Missense Mutation | ACC,ATC | T,I 14 | NP_620118.1 | |
NM_138764.4 | 110 | Missense Mutation | ACC,ATC | T,I 14 | NP_620119.2 | |
XM_017027077.1 | 110 | UTR 5 | XP_016882566.1 |
FTL - ferritin light chain | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |