Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTTCAGGGTCAGAGCCTGAGTCCT[C/T]GCCTTGTTGGTTCTGAATCTGACTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 126391 MIM: 603602 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LIG1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LIG1 - DNA ligase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000234.2 | 2674 | Missense Mutation | AAG,GAG | K,E 909 | NP_000225.1 | |
NM_001289063.1 | 2674 | Missense Mutation | AAG,GAG | K,E 878 | NP_001275992.1 | |
NM_001289064.1 | 2674 | Missense Mutation | AAG,GAG | K,E 841 | NP_001275993.1 | |
NM_001320970.1 | 2674 | Missense Mutation | AAG,GAG | K,E 908 | NP_001307899.1 | |
NM_001320971.1 | 2674 | Missense Mutation | AAG,GAG | K,E 879 | NP_001307900.1 | |
XM_017026809.1 | 2674 | Missense Mutation | AAG,GAG | K,E 873 | XP_016882298.1 |
PLA2G4C - phospholipase A2 group IVC | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |