Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGTCATCCACTCACAGGAGCAAGA[A/G]GTGCTCAGGAGCACCTTTGTGTCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
THEG5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
THEG5 - testis highly expressed protein 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001205273.2 | Intron | NP_001192202.1 | ||||
NM_001278575.1 | Intron | NP_001265504.1 | ||||
NM_001278576.1 | Intron | NP_001265505.1 | ||||
NM_001278577.1 | Intron | NP_001265506.1 | ||||
NM_001278578.1 | Intron | NP_001265507.1 | ||||
XM_011526302.2 | Intron | XP_011524604.1 | ||||
XM_011526303.2 | Intron | XP_011524605.1 | ||||
XM_017026126.1 | Intron | XP_016881615.1 | ||||
XM_017026127.1 | Intron | XP_016881616.1 | ||||
XM_017026128.1 | Intron | XP_016881617.1 | ||||
XM_017026129.1 | Intron | XP_016881618.1 | ||||
XM_017026130.1 | Intron | XP_016881619.1 | ||||
XM_017026131.1 | Intron | XP_016881620.1 | ||||
XM_017026132.1 | Intron | XP_016881621.1 | ||||
XM_017026133.1 | Intron | XP_016881622.1 |