Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTCCCTGGCGGCCCGTGGCCTCTC[C/G]TCTTTCCGCGGCCTCTCTTTCTTAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600957 MIM: 608743 MIM: 600796 | ||||||||||||||||||||
Literature Links: |
AMH PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
AMH - anti-Mullerian hormone | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
JSRP1 - junctional sarcoplasmic reticulum protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_144616.3 | 1278 | Silent Mutation | GAC,GAG | D,E 264 | NP_653217.1 | |
XM_017026285.1 | 1278 | Missense Mutation | GAC,GAG | D,E 317 | XP_016881774.1 | |
XM_017026286.1 | 1278 | Missense Mutation | GAC,GAG | D,E 310 | XP_016881775.1 | |
XM_017026287.1 | 1278 | Missense Mutation | GAC,GAG | D,E 271 | XP_016881776.1 |
MIR4321 - microRNA 4321 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SF3A2 - splicing factor 3a subunit 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |