Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTGAGGATGAGTTGCTCTCCCAGG[C/T]CCTCCTGGGCGCCAAGAAGCCTATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 132880 MIM: 610675 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NR2F6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NR2F6 - nuclear receptor subfamily 2 group F member 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
OCEL1 - occludin/ELL domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024578.2 | 415 | Missense Mutation | GCC,GTC | A,V 124 | NP_078854.1 | |
XM_005260079.2 | 415 | Missense Mutation | GCC,GTC | A,V 68 | XP_005260136.1 | |
XM_006722899.3 | 415 | Missense Mutation | GCC,GTC | A,V 124 | XP_006722962.1 | |
XM_017027306.1 | 415 | Missense Mutation | GCC,GTC | A,V 68 | XP_016882795.1 |
USE1 - unconventional SNARE in the ER 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |