Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGACTCAGTAAGTGAAGGAAATGCC[A/G]GGCTTTATCGCTGCCTCTATTATAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602993 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LAIR2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LAIR2 - leukocyte associated immunoglobulin like receptor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002288.5 | 411 | Missense Mutation | AGG,GGG | R,G 97 | NP_002279.2 | |
NM_021270.4 | 411 | Missense Mutation | AGG,GGG | R,G 97 | NP_067154.1 | |
XM_011526961.2 | 411 | Missense Mutation | AGG,GGG | R,G 85 | XP_011525263.1 |