Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAGCTTGCAGGGAAATCAGTTATA[C/T]GAGTTTCCATCTGTTCACACATTTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF233 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF233 - zinc finger protein 233 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001207005.1 | Intron | NP_001193934.1 | ||||
NM_181756.2 | Intron | NP_861421.2 | ||||
XM_017026758.1 | Intron | XP_016882247.1 | ||||
XM_017026759.1 | Intron | XP_016882248.1 | ||||
XM_017026760.1 | Intron | XP_016882249.1 | ||||
XM_017026761.1 | Intron | XP_016882250.1 |