Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCGCATGGCTGGATTTACCCTGGCC[C/T]AGTAGTTCATGGCTACTGGTTCCGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605640 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SIGLEC9 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SIGLEC9 - sialic acid binding Ig like lectin 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001198558.1 | 231 | Missense Mutation | CCA,CTA | P,L 55 | NP_001185487.1 | |
NM_014441.2 | 231 | Missense Mutation | CCA,CTA | P,L 55 | NP_055256.1 | |
XM_006723146.3 | 231 | Missense Mutation | CCA,CTA | P,L 55 | XP_006723209.1 | |
XM_011526730.2 | 231 | Missense Mutation | CCA,CTA | P,L 55 | XP_011525032.1 | |
XM_011526732.2 | 231 | Missense Mutation | CCA,CTA | P,L 55 | XP_011525034.1 | |
XM_017026595.1 | 231 | Missense Mutation | CCA,CTA | P,L 55 | XP_016882084.1 | |
XM_017026596.1 | 231 | Missense Mutation | CCA,CTA | P,L 55 | XP_016882085.1 |