Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGAACCCTGCCAAGAAGTCGGGCT[C/G]TGGCTGGGCCTCTGTCAAGTACAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607881 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MPND PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MPND - MPN domain containing | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001159846.2 | 495 | Missense Mutation | TCT,TGT | S,C 143 | NP_001153318.1 | |
NM_001300862.1 | 495 | Missense Mutation | TCT,TGT | S,C 143 | NP_001287791.1 | |
NM_032868.5 | 495 | Missense Mutation | TCT,TGT | S,C 143 | NP_116257.2 | |
XM_006722926.2 | 495 | Missense Mutation | TCT,TGT | S,C 143 | XP_006722989.1 |
STAP2 - signal transducing adaptor family member 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |