Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGCGCCCCCCGACAGGCCCACGCCC[G/T]CCAGGCCGCTCATACCATCGATGGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603458 MIM: 177060 | ||||||||||||||||||||
Literature Links: |
ELAVL3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ELAVL3 - ELAV like neuron-specific RNA binding protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001420.3 | 1195 | Missense Mutation | GAG,GCG | E,A 271 | NP_001411.2 | |
NM_032281.2 | 1195 | Missense Mutation | GAG,GCG | E,A 264 | NP_115657.2 | |
XM_011527778.2 | 1195 | Missense Mutation | GAG,GCG | E,A 270 | XP_011526080.1 |
PRKCSH - protein kinase C substrate 80K-H | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |