Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATGACCTCCATGGGTCATTACCTC[C/T]ATCCCTCTGCCTCTAGGAGCCACCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604520 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TNFSF14 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TNFSF14 - tumor necrosis factor superfamily member 14 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003807.4 | 3033 | UTR 3 | NP_003798.2 | |||
NM_172014.3 | 3033 | UTR 3 | NP_742011.2 | |||
XM_017027417.1 | 3033 | UTR 3 | XP_016882906.1 | |||
XM_017027418.1 | 3033 | Intron | XP_016882907.1 |