Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCCCATCATGAGAGCGGGCCACAG[C/T]GTTGATATCAGTGCCGTCCGCCCCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
EML2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
EML2 - echinoderm microtubule associated protein like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001193268.1 | 2489 | Missense Mutation | NP_001180197.1 | |||
NM_001193269.1 | 2489 | Missense Mutation | NP_001180198.1 | |||
NM_012155.2 | 2489 | Missense Mutation | NP_036287.1 | |||
XM_006723125.2 | 2489 | Missense Mutation | XP_006723188.1 | |||
XM_006723126.2 | 2489 | Missense Mutation | XP_006723189.1 | |||
XM_011526694.2 | 2489 | Missense Mutation | XP_011524996.1 | |||
XM_011526695.2 | 2489 | Missense Mutation | XP_011524997.1 | |||
XM_011526698.2 | 2489 | Missense Mutation | XP_011525000.1 | |||
XM_011526700.2 | 2489 | Missense Mutation | XP_011525002.1 | |||
XM_011526701.2 | 2489 | Missense Mutation | XP_011525003.1 | |||
XM_017026550.1 | 2489 | Missense Mutation | XP_016882039.1 | |||
XM_017026551.1 | 2489 | Intron | XP_016882040.1 |