Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCACATCCCACAGGGGCAGAAATCC[C/T]GCTCGAGAAGAGCTGAATTCGAAAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 120216 | ||||||||||||||||||||
Literature Links: |
COL5A3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
COL5A3 - collagen type V alpha 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015719.3 | 2929 | Silent Mutation | GCA,GCG | A,A 1714 | NP_056534.2 | |
XM_011528042.2 | 2929 | Silent Mutation | GCA,GCG | A,A 1713 | XP_011526344.1 | |
XM_017026849.1 | 2929 | Silent Mutation | GCA,GCG | A,A 935 | XP_016882338.1 |