Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCCTGGGTGGCTGGGAAGGTGGCC[C/T]GCCGGAGGGTGGGTGCCACCTGGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609949 MIM: 615475 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C5AR2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C5AR2 - complement component 5a receptor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271749.1 | 334 | Missense Mutation | CGC,TGC | R,C 65 | NP_001258678.1 | |
NM_001271750.1 | 334 | Missense Mutation | CGC,TGC | R,C 65 | NP_001258679.1 | |
NM_018485.2 | 334 | Missense Mutation | CGC,TGC | R,C 65 | NP_060955.1 | |
XM_011526736.2 | 334 | Missense Mutation | CGC,TGC | R,C 65 | XP_011525038.1 |
DHX34 - DEAH-box helicase 34 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |