Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCATTTATTGTTCAGGGGCTGAGCC[C/T]CCCTGTTTTGGGGGGGTTCCTGCAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608253 | ||||||||||||||||||||
Literature Links: |
TSKS PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TSKS - testis specific serine kinase substrate | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021733.1 | 1769 | Silent Mutation | GGA,GGG | G,G 586 | NP_068379.1 | |
XM_011527201.1 | 1769 | Silent Mutation | GGA,GGG | G,G 611 | XP_011525503.1 | |
XM_011527202.1 | 1769 | UTR 3 | XP_011525504.1 | |||
XM_011527203.2 | 1769 | UTR 3 | XP_011525505.1 |