Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCTGTAATACTGACAGCCATATCG[C/T]CCTGTGTGTTCCCAGGTGATGGATG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604815 | ||||||||||||||||||||
Literature Links: |
LILRB2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LILRB2 - leukocyte immunoglobulin like receptor B2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080978.3 | Intron | NP_001074447.2 | ||||
NM_001278403.2 | Intron | NP_001265332.2 | ||||
NM_001278404.2 | Intron | NP_001265333.2 | ||||
NM_001278405.2 | Intron | NP_001265334.2 | ||||
NM_001278406.2 | Intron | NP_001265335.2 | ||||
NM_005874.4 | Intron | NP_005865.3 |
MIR4752 - microRNA 4752 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |