Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTGGGGGCGCTCCGGCTGCCGTAC[C/T]GTGGCTGCAAGCCTAAACCGGGCTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605312 MIM: 607518 | ||||||||||||||||||||
Literature Links: |
GDF15 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GDF15 - growth differentiation factor 15 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LRRC25 - leucine rich repeat containing 25 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145256.2 | 855 | Missense Mutation | NP_660299.2 | |||
XM_005259739.3 | 855 | Missense Mutation | XP_005259796.1 |
MIR3189 - microRNA 3189 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |