Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTAGCTGCTCCAGGCTGAGCAGGGA[C/T]AGGCCCAGCTGATCCTCCTGCAGAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 126340 MIM: 601334 | ||||||||||||||||||||
Literature Links: |
ERCC2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ERCC2 - ERCC excision repair 2, TFIIH core complex helicase subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000400.3 | 2237 | Silent Mutation | CTA,CTG | L,L 736 | NP_000391.1 | |
NM_001130867.1 | 2237 | Intron | NP_001124339.1 | |||
XM_011526611.2 | 2237 | Silent Mutation | CTA,CTG | L,L 710 | XP_011524913.1 | |
XM_017026467.1 | 2237 | Silent Mutation | CTA,CTG | L,L 695 | XP_016881956.1 |
KLC3 - kinesin light chain 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_177417.2 | 2237 | Intron | NP_803136.2 |