Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACCCCACCTGGCGAGAAGGTGGAT[A/G]TACCCGCAGGCCAGGGCAGCCCCTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ABHD17A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ABHD17A - abhydrolase domain containing 17A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130111.1 | 3746 | Intron | NP_001123583.1 | |||
NM_031213.3 | 3746 | Intron | NP_112490.3 | |||
XM_011528338.1 | 3746 | Silent Mutation | TAC,TAT | Y,Y 156 | XP_011526640.1 | |
XM_017027341.1 | 3746 | Intron | XP_016882830.1 | |||
XM_017027342.1 | 3746 | Intron | XP_016882831.1 |