Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGGCCGGACGGCCACGTGTGGCTG[C/G]GTCTCGGTATAGCCCCCGATGGGCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
MEIS3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MEIS3 - Meis homeobox 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001009813.2 | 1695 | Intron | NP_001009813.1 | |||
NM_001301059.1 | 1695 | Intron | NP_001287988.1 | |||
NM_020160.2 | 1695 | Intron | NP_064545.1 | |||
XM_011527136.2 | 1695 | Silent Mutation | ACC,ACG | T,T 421 | XP_011525438.1 | |
XM_011527138.2 | 1695 | Silent Mutation | ACC,ACG | T,T 371 | XP_011525440.1 | |
XM_011527139.2 | 1695 | Intron | XP_011525441.1 | |||
XM_011527140.1 | 1695 | Silent Mutation | ACC,ACG | T,T 218 | XP_011525442.1 | |
XM_017027011.1 | 1695 | Silent Mutation | ACC,ACG | T,T 438 | XP_016882500.1 |