Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAAAGACAACTTTTCTTGGAGCCA[A/G]GCCTCAGTGAGGTCATCAGTGGTGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604104 MIM: 612807 | ||||||||||||||||||||
Literature Links: |
GMFG PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GMFG - glia maturation factor gamma | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301008.1 | 506 | Silent Mutation | GCC,GCT | A,A 91 | NP_001287937.1 | |
NM_004877.3 | 506 | Silent Mutation | GCC,GCT | A,A 132 | NP_004868.1 |
LRFN1 - leucine rich repeat and fibronectin type III domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |