Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTCTGTGGGCCGGCTTCATCTCTC[G/T]GTGGCTTCCCTGTGGCTTCCGCAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611340 | ||||||||||||||||||||
Literature Links: |
ATG4D PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ATG4D - autophagy related 4D cysteine peptidase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001281504.1 | 1844 | Intron | NP_001268433.1 | |||
NM_032885.5 | 1844 | Intron | NP_116274.3 | |||
XM_006722927.1 | 1844 | Intron | XP_006722990.1 |
KRI1 - KRI1 homolog | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_023008.3 | 1844 | Missense Mutation | CAG,CCG | Q,P 612 | NP_075384.3 | |
XM_011528190.2 | 1844 | Missense Mutation | CAG,CCG | Q,P 494 | XP_011526492.1 |
MIR1238 - microRNA 1238 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |