Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAGGCTCACGGGGTGTCAGCTGAA[A/G]AAGTAGGTGGAACGCTTCACCTGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602781 MIM: 601778 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HCN2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HCN2 - hyperpolarization activated cyclic nucleotide gated potassium channel 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001194.3 | Intron | NP_001185.3 |
POLRMT - RNA polymerase mitochondrial | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005035.3 | Intron | NP_005026.3 | ||||
XM_005259580.3 | Intron | XP_005259637.2 |