Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTCCTAGTAGCGGCCGACCTTGGC[A/G]TCCCCAATGGCGCCCCAGACGTCAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605072 MIM: 601032 MIM: 176802 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GIPC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GIPC1 - GIPC PDZ domain containing family member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005716.3 | 894 | Silent Mutation | GAC,GAT | D,D 327 | NP_005707.1 | |
NM_202468.2 | 894 | Silent Mutation | GAC,GAT | D,D 327 | NP_974197.1 | |
NM_202469.2 | 894 | Silent Mutation | GAC,GAT | D,D 230 | NP_974198.1 | |
NM_202470.2 | 894 | Silent Mutation | GAC,GAT | D,D 327 | NP_974199.1 | |
NM_202494.2 | 894 | Silent Mutation | GAC,GAT | D,D 230 | NP_974223.1 | |
XM_017026147.1 | 894 | Silent Mutation | GAC,GAT | D,D 327 | XP_016881636.1 | |
XM_017026148.1 | 894 | Silent Mutation | GAC,GAT | D,D 230 | XP_016881637.1 |
PKN1 - protein kinase N1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PTGER1 - prostaglandin E receptor 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |