Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGCAGCCTCATCGCGGAAGGACT[C/T]AGCACAGCCTTGCAGCTGTTTGATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610197 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MED25 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MED25 - mediator complex subunit 25 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_030973.3 | 417 | Silent Mutation | CTC,CTT | L,L 118 | NP_112235.2 | |
XM_011527353.1 | 417 | Silent Mutation | CTC,CTT | L,L 118 | XP_011525655.1 |
MIR6800 - microRNA 6800 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |