Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCCTATTGCGCTGCTCAGCCCCAT[A/G]GCCAGCAACATCAGGAAGAAGATAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607972 | ||||||||||||||||||||
Literature Links: |
SLC6A16 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SLC6A16 - solute carrier family 6 member 16 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014037.2 | 1771 | Silent Mutation | GCC,GCT | A,A 507 | NP_054756.2 | |
XM_005258820.2 | 1771 | Silent Mutation | GCC,GCT | A,A 507 | XP_005258877.1 | |
XM_006723168.3 | 1771 | Silent Mutation | GCC,GCT | A,A 507 | XP_006723231.1 | |
XM_011526859.2 | 1771 | Silent Mutation | GCC,GCT | A,A 552 | XP_011525161.2 | |
XM_011526860.2 | 1771 | Silent Mutation | GCC,GCT | A,A 529 | XP_011525162.1 | |
XM_011526861.2 | 1771 | Silent Mutation | GCC,GCT | A,A 360 | XP_011525163.1 | |
XM_017026712.1 | 1771 | Silent Mutation | GCC,GCT | A,A 589 | XP_016882201.1 |