Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGGCAGAAGGATGCCTTCTCACCT[C/T]GGTTCAGGGTGGGGGTGCTGTTGAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616416 | ||||||||||||||||||||
Literature Links: |
ADGRL1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ADGRL1 - adhesion G protein-coupled receptor L1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001008701.2 | 3870 | Missense Mutation | CAA,CGA | Q,R 1178 | NP_001008701.1 | |
NM_014921.4 | 3870 | Missense Mutation | CAA,CGA | Q,R 1173 | NP_055736.2 | |
XM_005259818.3 | 3870 | Missense Mutation | CAA,CGA | Q,R 1173 | XP_005259875.1 | |
XM_011527796.2 | 3870 | Missense Mutation | CAA,CGA | Q,R 1185 | XP_011526098.1 | |
XM_011527798.2 | 3870 | Missense Mutation | CAA,CGA | Q,R 1180 | XP_011526100.1 | |
XM_011527801.2 | 3870 | Missense Mutation | CAA,CGA | Q,R 1105 | XP_011526103.1 | |
XM_017026475.1 | 3870 | Missense Mutation | CAA,CGA | Q,R 1186 | XP_016881964.1 | |
XM_017026476.1 | 3870 | Missense Mutation | CAA,CGA | Q,R 1186 | XP_016881965.1 | |
XM_017026477.1 | 3870 | Missense Mutation | CAA,CGA | Q,R 1179 | XP_016881966.1 | |
XM_017026478.1 | 3870 | Missense Mutation | CAA,CGA | Q,R 1186 | XP_016881967.1 | |
XM_017026479.1 | 3870 | Missense Mutation | CAA,CGA | Q,R 1186 | XP_016881968.1 | |
XM_017026480.1 | 3870 | Missense Mutation | CAA,CGA | Q,R 1100 | XP_016881969.1 | |
XM_017026481.1 | 3870 | Intron | XP_016881970.1 |
LOC100507373 - uncharacterized LOC100507373 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |