Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCCGTCAGGGGCCGGAAGGACTGG[C/G]CCCGGTTGTCCAGTACAAACAGAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603314 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC27A5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC27A5 - solute carrier family 27 member 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321196.1 | 2116 | Missense Mutation | GCC,GGC | A,G 585 | NP_001308125.1 | |
NM_012254.2 | 2116 | Missense Mutation | GCC,GGC | A,G 669 | NP_036386.1 | |
XM_011526364.1 | 2116 | UTR 3 | XP_011524666.1 | |||
XM_017026214.1 | 2116 | Intron | XP_016881703.1 |
ZNF446 - zinc finger protein 446 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |